Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 134951247-134951416 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHB1
Entrez Gene ID
2047
Gene Name
EPH receptor B1
Gene Aliases
ELK, EPHT2, Hek6, NET
UniGene
Hs.116092
Gene Chromosome Location
Chr.3: 134776089-135260467 on Build GRCh38
Transcripts
NM_004441.4 XM_017005866.1 XM_017005867.1 XM_017005868.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6859380 c.130G>T p.E44* Chr.3: 134951377-134951377 on Build GRCh38
6983519 c.145G>A p.D49N Chr.3: 134951392-134951392 on Build GRCh38
5746188 c.124T>C p.W42R Chr.3: 134951371-134951371 on Build GRCh38
353176 c.153C>T p.N51N Chr.3: 134951400-134951400 on Build GRCh38
166041 c.149delA p.N51fs*2 Chr.3: 134951396-134951396 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770655335 Chr.3: 134951347-134951347 on Build GRCh38
rs574277818 Chr.3: 134951273-134951273 on Build GRCh38
rs371726436 Chr.3: 134951391-134951391 on Build GRCh38
rs201609946 Chr.3: 134951326-134951326 on Build GRCh38
rs577075709 Chr.3: 134951361-134951361 on Build GRCh38
rs774080528 Chr.3: 134951352-134951352 on Build GRCh38
rs751458110 Chr.3: 134951333-134951334 on Build GRCh38
rs747607901 Chr.3: 134951328-134951328 on Build GRCh38
rs367623340 Chr.3: 134951331-134951331 on Build GRCh38
rs764139128 Chr.3: 134951392-134951392 on Build GRCh38
rs985758591 Chr.3: 134951269-134951269 on Build GRCh38
rs74776697 Chr.3: 134951320-134951320 on Build GRCh38
rs771963295 Chr.3: 134951367-134951367 on Build GRCh38
rs965581992 Chr.3: 134951299-134951299 on Build GRCh38
rs536887246 Chr.3: 134951274-134951274 on Build GRCh38
rs777463411 Chr.3: 134951341-134951341 on Build GRCh38

View Full Product Details