Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44886339-44886464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGB2
Entrez Gene ID
3689
Gene Name
integrin subunit beta 2
Gene Aliases
CD18, LAD, LCAMB, LFA-1, MAC-1, MF17, MFI7
UniGene
Hs.375957
Gene Chromosome Location
Chr.21: 44885949-44928873 on Build GRCh38
Transcripts
NM_000211.4 NM_001127491.2 NM_001303238.1 XM_006724001.1 XM_017028341.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7334360 c.2273C>A p.T758N Chr.21: 44886405-44886405 on Build GRCh38
5807517 c.2253T>C p.N751N Chr.21: 44886425-44886425 on Build GRCh38
189199 c.2268C>T p.S756S Chr.21: 44886410-44886410 on Build GRCh38
2821422 c.2275A>G p.T759A Chr.21: 44886403-44886403 on Build GRCh38
7149089 c.2262C>T p.F754F Chr.21: 44886416-44886416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200448015 Chr.21: 44886401-44886401 on Build GRCh38
rs745761943 Chr.21: 44886362-44886362 on Build GRCh38
rs769739259 Chr.21: 44886444-44886444 on Build GRCh38
rs774665195 Chr.21: 44886374-44886374 on Build GRCh38
rs780740229 Chr.21: 44886437-44886437 on Build GRCh38
rs750477817 Chr.21: 44886383-44886383 on Build GRCh38
rs542061261 Chr.21: 44886399-44886399 on Build GRCh38
rs764387110 Chr.21: 44886372-44886372 on Build GRCh38
rs762881198 Chr.21: 44886368-44886368 on Build GRCh38
rs984333477 Chr.21: 44886365-44886365 on Build GRCh38
rs145385843 Chr.21: 44886398-44886398 on Build GRCh38
rs192197684 Chr.21: 44886395-44886395 on Build GRCh38
rs375133952 Chr.21: 44886361-44886361 on Build GRCh38
rs368180990 Chr.21: 44886367-44886367 on Build GRCh38
rs750065651 Chr.21: 44886443-44886443 on Build GRCh38
rs375931424 Chr.21: 44886410-44886410 on Build GRCh38
rs1010831564 Chr.21: 44886379-44886379 on Build GRCh38
rs572463215 Chr.21: 44886414-44886414 on Build GRCh38
rs372064061 Chr.21: 44886376-44886376 on Build GRCh38

View Full Product Details