Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC22A6
Entrez Gene ID
9356
Gene Name
solute carrier family 22 member 6
Gene Aliases
HOAT1, OAT1, PAHT, ROAT1
Chromosome Location
Chr.11: 62976597 - 62985023 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004790.4 NP_004781.2 1 - 2 702 79
NM_153276.2 NP_695008.1 1 - 2 702 79
NM_153277.2 NP_695009.1 1 - 2 702 79
NM_153278.2 NP_695010.1 1 - 2 702 79
XM_017018562.1 XP_016874051.1 1 - 2 650 79
GenBank mRNA AB009697.1 - 1 - 2 636 79
AB009698.1 - 1 - 2 633 79
AF057039.2 - 1 - 2 613 79
AF097490.1 - 1 - 2 369 79
AF104038.1 - 1 - 2 610 79
AF124373.1 - 1 - 2 626 79
AJ251529.1 - 1 - 2 369 79
AJ271205.1 - 1 - 2 369 79
AK055764.1 - 2 - 3 615 79
AK091879.1 - 1 - 2 627 79
AK298374.1 - 2 - 3 512 79
BC033682.1 - 1 - 2 662 79

View Full Product Details