Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 48431206-48431366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP22
Entrez Gene ID
58504
Gene Name
Rho GTPase activating protein 22
Gene Aliases
RhoGAP2, RhoGap22
UniGene
Hs.655672
Gene Chromosome Location
Chr.10: 48429831-48656265 on Build GRCh38
Transcripts
XM_011540013.2 XM_017016473.1 XM_017016476.1 NM_001278547.1 NM_001278548.1 NM_001323302.1 NM_001323320.1 NM_001323321.1 NM_001323322.1 NM_001323323.1 NM_001323324.1 NM_001323325.1 NM_001323326.1 NM_001323327.1 NM_001323328.1 NM_001323329.1 NM_001323330.1 NM_001323331.1 NM_139046.3 NM_139049.3 NR_136583.1 NR_136584.1 NR_136585.1

Gene Symbol
MAPK8
Entrez Gene ID
5599
Gene Name
mitogen-activated protein kinase 8
Gene Aliases
JNK, JNK-46, JNK1, JNK1A2, JNK21B1/2, PRKM8, SAPK1, SAPK1c
UniGene
Hs.138211
Gene Chromosome Location
Chr.10: 48306639-48439360 on Build GRCh38
Transcripts
XM_011540013.2 XM_017016473.1 XM_017016476.1 NM_001278547.1 NM_001278548.1 NM_001323302.1 NM_001323320.1 NM_001323321.1 NM_001323322.1 NM_001323323.1 NM_001323324.1 NM_001323325.1 NM_001323326.1 NM_001323327.1 NM_001323328.1 NM_001323329.1 NM_001323330.1 NM_001323331.1 NM_139046.3 NM_139049.3 NR_136583.1 NR_136584.1 NR_136585.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1686613 c.1138+2T>C p.? Chr.10: 48431272-48431272 on Build GRCh38
6189488 c.1109G>C p.G370A Chr.10: 48431241-48431241 on Build GRCh38
6189489 c.1109G>C p.G370A Chr.10: 48431241-48431241 on Build GRCh38
1686612 c.1138+2T>C p.? Chr.10: 48431272-48431272 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759471301 Chr.10: 48431305-48431309 on Build GRCh38
rs747259337 Chr.10: 48431288-48431288 on Build GRCh38
rs752334346 Chr.10: 48431249-48431249 on Build GRCh38
rs28395809 Chr.10: 48431231-48431231 on Build GRCh38
rs772091449 Chr.10: 48431281-48431281 on Build GRCh38
rs531142189 Chr.10: 48431309-48431309 on Build GRCh38
rs767178901 Chr.10: 48431246-48431246 on Build GRCh38
rs151285118 Chr.10: 48431253-48431253 on Build GRCh38
rs751150249 Chr.10: 48431232-48431232 on Build GRCh38
rs549255000 Chr.10: 48431329-48431329 on Build GRCh38
rs563677532 Chr.10: 48431302-48431302 on Build GRCh38
rs778058488 Chr.10: 48431252-48431252 on Build GRCh38
rs778785038 Chr.10: 48431323-48431323 on Build GRCh38
rs757766119 Chr.10: 48431254-48431254 on Build GRCh38
rs774483305 Chr.10: 48431311-48431311 on Build GRCh38
rs778924369 Chr.10: 48431261-48431261 on Build GRCh38
rs988262121 Chr.10: 48431247-48431247 on Build GRCh38
rs201573286 Chr.10: 48431233-48431233 on Build GRCh38
rs745968638 Chr.10: 48431265-48431265 on Build GRCh38
rs375812588 Chr.10: 48431296-48431296 on Build GRCh38
rs756498386 Chr.10: 48431250-48431250 on Build GRCh38
rs769615460 Chr.10: 48431294-48431294 on Build GRCh38
rs775748147 Chr.10: 48431283-48431283 on Build GRCh38

View Full Product Details