Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 134925739-134925907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHB1
Entrez Gene ID
2047
Gene Name
EPH receptor B1
Gene Aliases
ELK, EPHT2, Hek6, NET
UniGene
Hs.116092
Gene Chromosome Location
Chr.3: 134776089-135260467 on Build GRCh38
Transcripts
NM_004441.4 XM_017005866.1 XM_017005867.1 XM_017005868.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5608559 c.123+1G>A p.? Chr.3: 134925881-134925881 on Build GRCh38
3122881 c.88G>A p.A30T Chr.3: 134925845-134925845 on Build GRCh38
1419249 c.117G>A p.A39A Chr.3: 134925874-134925874 on Build GRCh38
3122879 c.81G>A p.T27T Chr.3: 134925838-134925838 on Build GRCh38
1419247 c.104C>T p.T35M Chr.3: 134925861-134925861 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867384092 Chr.3: 134925840-134925840 on Build GRCh38
rs767609694 Chr.3: 134925809-134925809 on Build GRCh38
rs764943074 Chr.3: 134925776-134925776 on Build GRCh38
rs756194825 Chr.3: 134925820-134925820 on Build GRCh38
rs539126781 Chr.3: 134925777-134925777 on Build GRCh38
rs779291109 Chr.3: 134925843-134925843 on Build GRCh38
rs1032625378 Chr.3: 134925764-134925764 on Build GRCh38
rs635721 Chr.3: 134925794-134925794 on Build GRCh38
rs375204964 Chr.3: 134925853-134925853 on Build GRCh38
rs556035012 Chr.3: 134925873-134925873 on Build GRCh38
rs75624920 Chr.3: 134925793-134925793 on Build GRCh38
rs762148161 Chr.3: 134925874-134925874 on Build GRCh38
rs376895227 Chr.3: 134925819-134925819 on Build GRCh38
rs772410007 Chr.3: 134925847-134925847 on Build GRCh38
rs754908715 Chr.3: 134925799-134925799 on Build GRCh38
rs773003001 Chr.3: 134925785-134925785 on Build GRCh38
rs536029159 Chr.3: 134925861-134925861 on Build GRCh38
rs779614389 Chr.3: 134925794-134925795 on Build GRCh38
rs759393707 Chr.3: 134925879-134925879 on Build GRCh38
rs769441012 Chr.3: 134925862-134925862 on Build GRCh38
rs113829630 Chr.3: 134925794-134925794 on Build GRCh38
rs368378750 Chr.3: 134925877-134925877 on Build GRCh38
rs370590244 Chr.3: 134925838-134925838 on Build GRCh38
rs773064927 Chr.3: 134925837-134925837 on Build GRCh38
rs772984614 Chr.3: 134925863-134925863 on Build GRCh38
rs777918167 Chr.3: 134925827-134925827 on Build GRCh38
rs746365113 Chr.3: 134925845-134925845 on Build GRCh38
rs762778248 Chr.3: 134925786-134925786 on Build GRCh38

View Full Product Details