Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 228296-228450 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SDHA
Entrez Gene ID
6389
Gene Name
succinate dehydrogenase complex flavoprotein subunit A
Gene Aliases
CMD1GG, FP, PGL5, SDH1, SDH2, SDHF
UniGene
Hs.440475
Gene Chromosome Location
Chr.5: 218223-264816 on Build GRCh38
Transcripts
NM_001294332.1 NM_004168.3 XM_005248331.3 XM_011514072.1 XM_011514073.1 XM_017009685.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1636684 c.748A>G p.K250E Chr.5: 228311-228311 on Build GRCh38
290530 c.743G>C p.R248T Chr.5: 228306-228306 on Build GRCh38
6773765 c.738C>T p.R246R Chr.5: 228301-228301 on Build GRCh38
6941006 c.733C>T p.H245Y Chr.5: 228296-228296 on Build GRCh38
7389029 c.760G>T p.V254F Chr.5: 228323-228323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998713774 Chr.5: 228412-228412 on Build GRCh38
rs770997653 Chr.5: 228340-228340 on Build GRCh38
rs749970496 Chr.5: 228383-228383 on Build GRCh38
rs771874095 Chr.5: 228390-228390 on Build GRCh38
rs368099030 Chr.5: 228372-228372 on Build GRCh38
rs756974060 Chr.5: 228378-228378 on Build GRCh38
rs1018316617 Chr.5: 228377-228377 on Build GRCh38
rs749566947 Chr.5: 228333-228333 on Build GRCh38
rs776403592 Chr.5: 228323-228323 on Build GRCh38
rs780311750 Chr.5: 228370-228373 on Build GRCh38
rs764755535 Chr.5: 228379-228379 on Build GRCh38
rs775912212 Chr.5: 228356-228356 on Build GRCh38
rs372913930 Chr.5: 228350-228354 on Build GRCh38
rs772227580 Chr.5: 228357-228358 on Build GRCh38
rs372293235 Chr.5: 228351-228351 on Build GRCh38
rs201245536 Chr.5: 228345-228345 on Build GRCh38
rs761233834 Chr.5: 228360-228360 on Build GRCh38
rs538621686 Chr.5: 228342-228342 on Build GRCh38
rs1041809852 Chr.5: 228324-228349 on Build GRCh38
rs1057517564 Chr.5: 228352-228356 on Build GRCh38
rs753318329 Chr.5: 228388-228388 on Build GRCh38
rs1126412 Chr.5: 228320-228320 on Build GRCh38
rs764620283 Chr.5: 228364-228364 on Build GRCh38

View Full Product Details