The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001195248.1 | NP_001182177.1 | 2 - 3 | 216 | 98 |
NM_001195249.1 | NP_001182178.1 | 2 - 3 | 385 | 98 | |
NM_001195251.1 | NP_001182180.1 | 3 - 4 | 322 | 98 | |
NM_001195252.1 | NP_001182181.1 | 2 - 3 | 216 | 98 | |
NM_175069.2 | NP_778239.1 | 2 - 3 | 216 | 98 | |
NM_175073.2 | NP_778243.1 | 3 - 4 | 322 | 98 | |
XM_006716791.3 | XP_006716854.1 | 2 - 3 | 224 | 98 | |
XM_017014831.1 | XP_016870320.1 | 2 - 3 | 216 | 98 | |
XM_017014832.1 | XP_016870321.1 | 2 - 3 | 216 | 98 | |
XM_017014833.1 | XP_016870322.1 | 2 - 3 | 216 | 98 | |
XM_017014834.1 | XP_016870323.1 | 2 - 3 | 216 | 98 | |
XM_017014835.1 | XP_016870324.1 | 2 - 3 | 216 | 98 | |
XM_017014836.1 | XP_016870325.1 | 2 - 3 | 216 | 98 | |
GenBank mRNA | AB603741.1 | - | 1 - 2 | 112 | 98 |
AJ565854.1 | - | 1 - 2 | 197 | 98 | |
AK055672.1 | - | 2 - 3 | 203 | 98 | |
AK131046.1 | - | 2 - 3 | 168 | 98 | |
AY040777.1 | - | 2 - 3 | 191 | 98 | |
AY208829.1 | - | 2 - 3 | 191 | 98 | |
AY208831.1 | - | 2 - 3 | 182 | 98 | |
AY208834.1 | - | 2 - 3 | 187 | 98 | |
AY208836.1 | - | 2 - 3 | 331 | 98 | |
AY208837.1 | - | 3 - 4 | 288 | 98 | |
AY302067.1 | - | 1 - 2 | 334 | 98 | |
AY302071.1 | - | 1 - 2 | 334 | 98 | |
BC068107.1 | - | 3 - 4 | 297 | 98 | |
BC104881.1 | - | 2 - 3 | 230 | 98 | |
BQ957536.1 | - | 3 - 4 | 305 | 98 | |
BX538161.1 | - | 3 - 4 | 301 | 98 |