Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 135106400-135106568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHB1
Entrez Gene ID
2047
Gene Name
EPH receptor B1
Gene Aliases
ELK, EPHT2, Hek6, NET
UniGene
Hs.116092
Gene Chromosome Location
Chr.3: 134776089-135260467 on Build GRCh38
Transcripts
NM_004441.4 XM_017005866.1 XM_017005867.1 XM_017005868.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6861136 c.869A>C p.N290T Chr.3: 135106511-135106511 on Build GRCh38
1661689 c.840A>C p.E280D Chr.3: 135106482-135106482 on Build GRCh38
6933836 c.914C>A p.T305N Chr.3: 135106556-135106556 on Build GRCh38
4636719 c.868A>C p.N290H Chr.3: 135106510-135106510 on Build GRCh38
6945701 c.902G>A p.C301Y Chr.3: 135106544-135106544 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757126567 Chr.3: 135106428-135106428 on Build GRCh38
rs869302984 Chr.3: 135106454-135106454 on Build GRCh38
rs747232184 Chr.3: 135106461-135106461 on Build GRCh38
rs199734379 Chr.3: 135106533-135106533 on Build GRCh38
rs201455572 Chr.3: 135106507-135106507 on Build GRCh38
rs140381340 Chr.3: 135106501-135106502 on Build GRCh38
rs770155104 Chr.3: 135106479-135106479 on Build GRCh38
rs765192484 Chr.3: 135106529-135106529 on Build GRCh38
rs959430781 Chr.3: 135106526-135106526 on Build GRCh38
rs758548012 Chr.3: 135106441-135106441 on Build GRCh38
rs749856804 Chr.3: 135106498-135106498 on Build GRCh38
rs780036502 Chr.3: 135106456-135106456 on Build GRCh38
rs1015764975 Chr.3: 135106439-135106439 on Build GRCh38
rs375467924 Chr.3: 135106427-135106427 on Build GRCh38
rs995446201 Chr.3: 135106516-135106516 on Build GRCh38
rs781506236 Chr.3: 135106475-135106475 on Build GRCh38
rs748442196 Chr.3: 135106476-135106476 on Build GRCh38
rs750444218 Chr.3: 135106435-135106435 on Build GRCh38
rs528866784 Chr.3: 135106503-135106503 on Build GRCh38
rs768130276 Chr.3: 135106511-135106511 on Build GRCh38
rs760267858 Chr.3: 135106508-135106508 on Build GRCh38
rs869275612 Chr.3: 135106452-135106452 on Build GRCh38
rs761687482 Chr.3: 135106517-135106517 on Build GRCh38
rs766360874 Chr.3: 135106546-135106546 on Build GRCh38
rs765210592 Chr.3: 135106433-135106433 on Build GRCh38
rs755103946 Chr.3: 135106470-135106470 on Build GRCh38
rs367853569 Chr.3: 135106494-135106494 on Build GRCh38
rs371303583 Chr.3: 135106512-135106512 on Build GRCh38

View Full Product Details