The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006260.4 | NP_006251.1 | 1 - 2 | 202 | 100 |
XM_011521104.2 | XP_011519406.1 | 1 - 2 | 196 | 100 | |
GenBank mRNA | AK292947.1 | - | 1 - 2 | 202 | 100 |
AK312861.1 | - | 1 - 2 | 193 | 100 | |
BC047936.1 | - | 1 - 2 | 184 | 100 |