The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006260.4 | NP_006251.1 | 3 - 4 | 439 | 127 |
XM_011521104.2 | XP_011519406.1 | 3 - 4 | 433 | 127 | |
XM_011521105.1 | XP_011519407.1 | 3 - 4 | 374 | 127 | |
XM_017020674.1 | XP_016876163.1 | 3 - 4 | 374 | 127 | |
GenBank mRNA | AK292947.1 | - | 3 - 4 | 439 | 127 |
AK312861.1 | - | 3 - 4 | 430 | 127 | |
BC047936.1 | - | 3 - 4 | 421 | 127 |