The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006260.4 | NP_006251.1 | 7 - 8 | 965 | 89 |
XM_011521104.2 | XP_011519406.1 | 8 - 9 | 1046 | 89 | |
XM_011521105.1 | XP_011519407.1 | 8 - 9 | 987 | 89 | |
XM_017020674.1 | XP_016876163.1 | 7 - 8 | 900 | 89 | |
XM_017020675.1 | XP_016876164.1 | 5 - 6 | 1473 | 89 | |
GenBank mRNA | AK292947.1 | - | 7 - 8 | 965 | 89 |
AK312861.1 | - | 8 - 9 | 956 | 89 | |
BC047936.1 | - | 7 - 8 | 947 | 89 |