Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TBC1D2B
Entrez Gene ID
23102
Gene Name
TBC1 domain family member 2B
Gene Aliases
-
Chromosome Location
Chr.15: 77994985 - 78077708 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015079.5 NP_055894.6 9 - 10 2270 80
NM_144572.1 NP_653173.1 9 - 10 2270 80
XM_011521387.2 XP_011519689.1 8 - 9 2215 80
XM_017022022.1 XP_016877511.1 11 - 12 2291 80
GenBank mRNA AB028978.1 - 9 - 10 2133 80
AK000173.1 - 2 - 3 1237 80
BC033712.1 - 4 - 5 1053 80
BC140766.1 - 8 - 9 1915 80
BC144482.1 - 8 - 9 1915 80
BC152465.1 - 9 - 10 2133 80

View Full Product Details