The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138644.2 | NP_619585.1 | 1 - 2 | 38 | 70 |
GenBank mRNA | AF295037.1 | - | 1 - 2 | 38 | 70 |