Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC44A3
Entrez Gene ID
126969
Gene Name
solute carrier family 44 member 3
Gene Aliases
CTL3
Chromosome Location
Chr.1: 94820342 - 94895247 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114106.2 NP_001107578.1 7 - 8 873 104
NM_001258340.1 NP_001245269.1 7 - 8 873 104
NM_001258341.1 NP_001245270.1 7 - 8 777 104
NM_001258342.1 NP_001245271.1 6 - 7 765 104
NM_001258343.1 NP_001245272.1 6 - 7 669 104
NM_001301079.1 NP_001288008.1 6 - 7 712 104
NM_152369.4 NP_689582.2 6 - 7 808 104
XM_005270440.2 XP_005270497.1 7 - 8 768 104
XM_005270441.2 XP_005270498.1 7 - 8 810 104
XM_005270442.1 XP_005270499.1 7 - 8 864 104
XM_011540632.2 XP_011538934.1 6 - 7 737 104
XM_011540633.2 XP_011538935.1 6 - 7 575 104
XM_011540635.2 XP_011538937.1 7 - 8 864 104
XM_011540636.2 XP_011538938.1 7 - 8 768 104
XM_017000262.1 XP_016855751.1 6 - 7 750 104
XM_017000263.1 XP_016855752.1 7 - 8 810 104
XM_017000264.1 XP_016855753.1 7 - 8 864 104
XM_017000265.1 XP_016855754.1 7 - 8 768 104
XM_017000266.1 XP_016855755.1 7 - 8 864 104
GenBank mRNA AK300193.1 - 6 - 7 606 104
AK301288.1 - 6 - 7 614 104
AK302381.1 - 4 - 5 457 104
AK303906.1 - 6 - 7 744 104
AK313574.1 - 7 - 8 761 104
AK316123.1 - 8 - 9 718 104
AL530322.3 - 7 - 8 865 104
AL540829.3 - 6 - 7 765 104
AY358659.1 - 3 - 4 327 104
BC033858.1 - 7 - 8 836 104
BC053877.1 - 6 - 7 731 104
BX648225.1 - 6 - 7 737 104

View Full Product Details