Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC44A3
Entrez Gene ID
126969
Gene Name
solute carrier family 44 member 3
Gene Aliases
CTL3
Chromosome Location
Chr.1: 94820342 - 94895247 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114106.2 NP_001107578.1 9 - 10 1179 96
NM_001258340.1 NP_001245269.1 9 - 10 1179 96
NM_001258341.1 NP_001245270.1 9 - 10 1083 96
NM_001258342.1 NP_001245271.1 8 - 9 1071 96
NM_001258343.1 NP_001245272.1 8 - 9 975 96
NM_001301079.1 NP_001288008.1 8 - 9 1018 96
NM_152369.4 NP_689582.2 8 - 9 1114 96
XM_005270440.2 XP_005270497.1 9 - 10 1074 96
XM_005270441.2 XP_005270498.1 9 - 10 1116 96
XM_005270442.1 XP_005270499.1 9 - 10 1170 96
XM_011540632.2 XP_011538934.1 8 - 9 1043 96
XM_011540633.2 XP_011538935.1 8 - 9 881 96
XM_011540635.2 XP_011538937.1 9 - 10 1170 96
XM_011540636.2 XP_011538938.1 9 - 10 1074 96
XM_017000262.1 XP_016855751.1 8 - 9 1056 96
XM_017000263.1 XP_016855752.1 9 - 10 1116 96
XM_017000264.1 XP_016855753.1 9 - 10 1170 96
XM_017000265.1 XP_016855754.1 9 - 10 1074 96
XM_017000266.1 XP_016855755.1 9 - 10 1170 96
GenBank mRNA AK300193.1 - 8 - 9 912 96
AK301288.1 - 8 - 10 96
AK302381.1 - 6 - 7 763 96
AK303906.1 - 8 - 9 1050 96
AK313574.1 - 9 - 10 1067 96
AK316123.1 - 10 - 11 1024 96
AY358659.1 - 5 - 6 633 96
BC033858.1 - 9 - 10 1142 96
BC053877.1 - 8 - 9 1037 96
BX648225.1 - 8 - 9 1043 96

View Full Product Details