The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205301.1 | NP_001192230.1 | 3 - 4 | 639 | 58 |
NM_152403.3 | NP_689616.2 | 3 - 4 | 639 | 58 | |
GenBank mRNA | AK096474.1 | - | 3 - 4 | 617 | 58 |
BC063822.1 | - | 3 - 4 | 480 | 58 | |
BX647551.1 | - | 3 - 4 | 489 | 58 |