The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286434.1 | NP_001273363.1 | 2 - 3 | 288 | 112 |
NM_153356.2 | NP_699187.1 | 3 - 4 | 396 | 112 | |
XM_006720409.3 | XP_006720472.1 | 3 - 4 | 426 | 112 | |
XM_006720410.3 | XP_006720473.1 | 2 - 3 | 318 | 112 | |
XM_011521281.2 | XP_011519583.1 | 3 - 4 | 425 | 112 | |
XM_011521282.2 | XP_011519584.1 | 3 - 4 | 425 | 112 | |
XM_011521283.2 | XP_011519585.1 | 3 - 4 | 425 | 112 | |
XM_011521284.2 | XP_011519586.1 | 2 - 3 | 205 | 112 | |
GenBank mRNA | AB449907.1 | - | 2 - 3 | 164 | 112 |
BC033516.1 | - | 3 - 4 | 355 | 112 |