The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153442.3 | NP_703143.1 | 2 - 3 | 830 | 56 |
GenBank mRNA | AJ505757.1 | - | 2 - 3 | 829 | 56 |
BC112011.1 | - | 2 - 3 | 827 | 56 | |
BC113462.1 | - | 2 - 3 | 815 | 56 |