Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL12RB1
Entrez Gene ID
3594
Gene Name
interleukin 12 receptor subunit beta 1
Gene Aliases
CD212, IL-12R-BETA1, IL12RB, IMD30
Chromosome Location
Chr.19: 18058994 - 18099027 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290023.1 NP_001276952.1 3 - 4 415 94
NM_001290024.1 NP_001276953.1 4 - 5 406 94
NM_005535.2 NP_005526.1 3 - 4 415 94
NM_153701.2 NP_714912.1 3 - 4 415 94
XM_006722741.3 XP_006722804.2 4 - 5 608 94
XM_011527966.2 XP_011526268.1 4 - 5 629 94
XM_011527967.2 XP_011526269.1 4 - 5 615 94
XM_011527968.2 XP_011526270.1 4 - 5 623 94
XM_011527969.2 XP_011526271.1 4 - 5 609 94
XM_011527970.2 XP_011526272.1 4 - 5 628 94
XM_011527971.2 XP_011526273.1 4 - 5 626 94
XM_011527972.2 XP_011526274.1 4 - 5 626 94
XM_011527973.2 XP_011526275.1 3 - 4 420 94
XM_011527974.2 XP_011526276.1 3 - 4 408 94
XM_011527975.2 XP_011526277.1 4 - 5 606 94
XM_011527976.2 XP_011526278.1 4 - 5 626 94
XM_011527977.2 XP_011526279.1 4 - 5 599 94
GenBank mRNA AK225919.1 - 3 - 4 346 94
AK290423.1 - 3 - 4 343 94
BC029121.1 - 3 - 4 304 94
BC137404.1 - 3 - 4 260 94
BC137406.1 - 3 - 4 260 94
BX647221.1 - 4 - 5 406 94
U03187.1 - 3 - 4 299 94

View Full Product Details