The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004473.3 | NP_004464.2 | 1 - 1 | 554 | 137 |
GenBank mRNA | U89995.1 | - | 1 - 1 | 582 | 137 |
X94553.1 | - | 1 - 1 | 137 |