The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005431.1 | NP_005422.1 | 1 - 2 | 126 | 106 |
GenBank mRNA | AF035587.1 | - | 1 - 2 | 126 | 106 |
AK313607.1 | - | 1 - 2 | 102 | 106 | |
BC042137.1 | - | 1 - 2 | 101 | 106 | |
Y08837.1 | - | 1 - 2 | 122 | 106 |