The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005472.4 | NP_005463.1 | 2 - 3 | 308 | 95 |
XM_017017047.1 | XP_016872536.1 | 3 - 4 | 378 | 95 | |
XM_017017048.1 | XP_016872537.1 | 4 - 5 | 474 | 95 | |
XM_017017049.1 | XP_016872538.1 | 2 - 3 | 324 | 95 | |
XM_017017050.1 | XP_016872539.1 | 2 - 3 | 3182 | 95 | |
XM_017017051.1 | XP_016872540.1 | 3 - 4 | 297 | 95 | |
XM_017017052.1 | XP_016872541.1 | 2 - 3 | 248 | 95 | |
GenBank mRNA | AK311876.1 | - | 1 - 2 | 200 | 95 |
BC110612.2 | - | 1 - 2 | 77 | 95 | |
BC113743.1 | - | 1 - 2 | 132 | 95 | |
CD709836.1 | - | 2 - 3 | 251 | 95 | |
CR936625.1 | - | 2 - 3 | 302 | 95 | |
DQ192291.1 | - | 2 - 3 | 302 | 95 | |
DQ192292.1 | - | 1 - 2 | 178 | 95 | |
DR006135.1 | - | 2 - 3 | 381 | 95 |