Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPRY3
Entrez Gene ID
10251
Gene Name
sprouty RTK signaling antagonist 3
Gene Aliases
spry-3
Chromosome Location
Chr.X: 155612565 - 155782459 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001304990.1 NP_001291919.1 2 - 3 259 95
NM_005840.2 NP_005831.1 1 - 2 403 95
GenBank mRNA AK125483.1 - 1 - 2 328 95
DA102958.1 - 2 - 3 259 95

View Full Product Details