The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016424.4 | NP_057508.2 | 10 - 10 | 2447 | 150 |
XM_005257448.1 | XP_005257505.1 | 11 - 11 | 2310 | 150 | |
XM_005257452.1 | XP_005257509.1 | 10 - 10 | 2169 | 150 | |
XM_006721943.2 | XP_006722006.1 | 11 - 11 | 2089 | 150 | |
GenBank mRNA | AF069250.1 | - | 1 - 1 | 1151 | 150 |
AK027723.1 | - | 1 - 1 | 892 | 150 | |
AL050121.1 | - | 1 - 1 | 783 | 150 |