The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032524.1 | NP_115913.1 | 1 - 1 | 743 | 152 |
GenBank mRNA | AJ296168.1 | - | 2 - 2 | 566 | 152 |
AJ406936.1 | - | 1 - 1 | 743 | 152 |