The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001201484.1 | NP_001188413.1 | 3 - 4 | 1007 | 101 |
NM_032634.3 | NP_116023.2 | 2 - 3 | 909 | 101 | |
NM_152850.3 | NP_690577.2 | 2 - 3 | 555 | 101 | |
XM_005251619.3 | XP_005251676.1 | 2 - 3 | 534 | 101 | |
XM_017015222.1 | XP_016870711.1 | 3 - 4 | 1026 | 101 | |
XM_017015223.1 | XP_016870712.1 | 2 - 3 | 936 | 101 | |
XM_017015224.1 | XP_016870713.1 | 2 - 3 | 1200 | 101 | |
GenBank mRNA | AK074064.1 | - | 2 - 3 | 530 | 101 |
AK090433.1 | - | 1 - 2 | 1498 | 101 | |
AL833956.1 | - | 2 - 3 | 945 | 101 | |
AY358472.1 | - | 1 - 2 | 515 | 101 | |
BC029271.1 | - | 1 - 2 | 180 | 101 | |
BC036916.1 | - | 1 - 2 | 520 | 101 | |
BC065282.1 | - | 3 - 4 | 678 | 101 | |
BI818120.1 | - | 2 - 3 | 549 | 101 | |
CD387014.1 | - | 2 - 3 | 546 | 101 | |
CR991745.1 | - | 2 - 3 | 555 | 101 |