The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001102610.1 | NP_001096080.1 | 5 - 6 | 613 | 85 |
NM_052903.4 | NP_443135.3 | 5 - 6 | 613 | 85 | |
XM_005272540.2 | XP_005272597.1 | 5 - 6 | 517 | 85 | |
XM_011543868.1 | XP_011542170.1 | 5 - 6 | 517 | 85 | |
XM_011543869.2 | XP_011542171.1 | 5 - 6 | 517 | 85 | |
XM_011543870.1 | XP_011542172.1 | 5 - 6 | 517 | 85 | |
XM_017021890.1 | XP_016877379.1 | 5 - 6 | 517 | 85 | |
XM_017021891.1 | XP_016877380.1 | 5 - 6 | 517 | 85 | |
XM_017021892.1 | XP_016877381.1 | 5 - 6 | 517 | 85 | |
XM_017021893.1 | XP_016877382.1 | 5 - 6 | 517 | 85 | |
XM_017021894.1 | XP_016877383.1 | 2 - 3 | 165 | 85 | |
XM_017021895.1 | XP_016877384.1 | 5 - 6 | 537 | 85 | |
XM_017021899.1 | XP_016877388.1 | 5 - 6 | 509 | 85 | |
XM_017021900.1 | XP_016877389.1 | 5 - 6 | 481 | 85 | |
GenBank mRNA | AB067486.2 | - | 5 - 6 | 527 | 85 |
AF272884.1 | - | 5 - 6 | 537 | 85 | |
AK056416.1 | - | 5 - 6 | 512 | 85 | |
AK289859.1 | - | 5 - 6 | 512 | 85 | |
AK310897.1 | - | 5 - 6 | 509 | 85 | |
BC071560.1 | - | 5 - 6 | 509 | 85 |