The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152527.4 | NP_689740.2 | 4 - 5 | 1839 | 106 |
XM_005246353.3 | XP_005246410.1 | 3 - 4 | 1695 | 106 | |
XM_011510751.2 | XP_011509053.1 | 4 - 5 | 1610 | 106 | |
XM_017003480.1 | XP_016858969.1 | 4 - 5 | 1533 | 106 | |
GenBank mRNA | AK055317.1 | - | 1 - 2 | 289 | 106 |
AK055356.1 | - | 2 - 3 | 1096 | 106 | |
AK292873.1 | - | 4 - 5 | 1571 | 106 | |
AL833470.1 | - | 1 - 2 | 338 | 106 | |
BC065524.1 | - | 4 - 5 | 1743 | 106 | |
BX648181.1 | - | 2 - 3 | 1007 | 106 |