The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022462.4 | NP_071907.4 | 11 - 12 | 1683 | 78 |
NM_152794.3 | NP_690007.1 | 12 - 13 | 1779 | 78 | |
NM_152795.3 | NP_690008.2 | 12 - 13 | 1747 | 78 | |
XM_005259152.4 | XP_005259209.1 | 13 - 14 | 1875 | 78 | |
XM_005259153.3 | XP_005259210.2 | 10 - 11 | 1546 | 78 | |
XM_017027132.1 | XP_016882621.1 | 13 - 14 | 2001 | 78 | |
XM_017027133.1 | XP_016882622.1 | 13 - 14 | 2001 | 78 | |
XM_017027134.1 | XP_016882623.1 | 12 - 13 | 1871 | 78 | |
XM_017027135.1 | XP_016882624.1 | 12 - 13 | 1905 | 78 | |
XM_017027136.1 | XP_016882625.1 | 13 - 14 | 1875 | 78 | |
XM_017027137.1 | XP_016882626.1 | 12 - 13 | 1905 | 78 | |
XM_017027138.1 | XP_016882627.1 | 12 - 13 | 1745 | 78 | |
XM_017027139.1 | XP_016882628.1 | 11 - 12 | 1795 | 78 | |
XM_017027140.1 | XP_016882629.1 | 11 - 12 | 1725 | 78 | |
XM_017027141.1 | XP_016882630.1 | 11 - 12 | 1767 | 78 | |
GenBank mRNA | AB054067.1 | - | 12 - 13 | 1710 | 78 |
AB295039.1 | - | 12 - 13 | 1728 | 78 | |
AF463492.1 | - | 12 - 13 | 1742 | 78 | |
AK021653.1 | - | 13 - 14 | 1773 | 78 | |
AK297828.1 | - | 11 - 12 | 1669 | 78 | |
AK299699.1 | - | 11 - 12 | 1593 | 78 | |
AK314680.1 | - | 12 - 13 | 1779 | 78 | |
BC080551.1 | - | 13 - 14 | 1718 | 78 |