The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258017.1 | NP_001244946.1 | 6 - 7 | 973 | 80 |
NM_001258018.1 | NP_001244947.1 | 6 - 7 | 1055 | 80 | |
NM_152899.1 | NP_690863.1 | 4 - 5 | 446 | 80 | |
NM_172374.2 | NP_758962.1 | 6 - 7 | 997 | 80 | |
NR_047577.1 | 7 - 8 | 1032 | 80 | ||
GenBank mRNA | AF293462.1 | - | 4 - 5 | 446 | 80 |
AJ880386.1 | - | 6 - 7 | 872 | 80 | |
AJ880387.1 | - | 6 - 7 | 849 | 80 | |
AJ880388.1 | - | 6 - 7 | 911 | 80 | |
AJ880389.1 | - | 7 - 8 | 909 | 80 | |
AY358933.1 | - | 4 - 5 | 446 | 80 | |
BC026103.1 | - | 6 - 7 | 901 | 80 | |
BC064378.1 | - | 4 - 5 | 432 | 80 | |
BC090852.1 | - | 4 - 5 | 433 | 80 | |
DQ079587.1 | - | 6 - 7 | 997 | 80 | |
DQ079588.1 | - | 6 - 7 | 1055 | 80 | |
DQ079589.1 | - | 6 - 7 | 973 | 80 |