The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278623.1 | NP_001265552.1 | 17 - 18 | 2138 | 65 |
NM_001278624.1 | NP_001265553.1 | 17 - 18 | 2276 | 65 | |
NM_152995.5 | NP_694540.3 | 17 - 18 | 2178 | 65 | |
GenBank mRNA | AB085695.1 | - | 17 - 18 | 2134 | 65 |
AB181916.1 | - | 17 - 18 | 2258 | 65 | |
AK131285.1 | - | 15 - 16 | 1724 | 65 | |
AK302440.1 | - | 17 - 18 | 2136 | 65 | |
AY134856.1 | - | 1 - 2 | 182 | 65 |