The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130045.1 | NP_001123517.1 | 7 - 8 | 783 | 70 |
NM_153254.2 | NP_694986.2 | 3 - 4 | 590 | 70 | |
XM_005244738.1 | XP_005244795.1 | 3 - 4 | 617 | 70 | |
XM_011541177.2 | XP_011539479.1 | 7 - 8 | 860 | 70 | |
XM_017000906.1 | XP_016856395.1 | 3 - 4 | 861 | 70 | |
XM_017000907.1 | XP_016856396.1 | 4 - 5 | 942 | 70 | |
XM_017000908.1 | XP_016856397.1 | 4 - 5 | 942 | 70 | |
XM_017000909.1 | XP_016856398.1 | 4 - 5 | 942 | 70 | |
XM_017000910.1 | XP_016856399.1 | 4 - 5 | 942 | 70 | |
XM_017000911.1 | XP_016856400.1 | 4 - 5 | 942 | 70 | |
XM_017000912.1 | XP_016856401.1 | 5 - 6 | 714 | 70 | |
GenBank mRNA | AK093438.1 | - | 3 - 4 | 590 | 70 |
BC027945.1 | - | 7 - 8 | 805 | 70 | |
BC126152.1 | - | 3 - 4 | 557 | 70 | |
BC126154.1 | - | 3 - 4 | 557 | 70 |