The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003807.4 | NP_003798.2 | 1 - 2 | 293 | 76 |
GenBank mRNA | AF064090.1 | - | 1 - 2 | 293 | 76 |