The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172198.2 | NP_751948.1 | 5 - 6 | 1944 | 64 |
XM_006710630.3 | XP_006710693.1 | 5 - 6 | 1609 | 64 | |
GenBank mRNA | AF048712.1 | - | 4 - 5 | 1464 | 64 |
AF187964.1 | - | 4 - 5 | 1536 | 64 | |
AF205856.1 | - | 4 - 5 | 1464 | 64 | |
BC113475.1 | - | 4 - 5 | 1486 | 64 | |
BC113477.1 | - | 4 - 5 | 1486 | 64 | |
BC143390.1 | - | 4 - 5 | 1486 | 64 |