The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033576.1 | NP_001028748.1 | 13 - 14 | 1924 | 61 |
NM_001267052.1 | NP_001253981.1 | 13 - 14 | 1921 | 61 | |
NM_001308281.1 | NP_001295210.1 | 10 - 11 | 1590 | 61 | |
NM_173167.3 | NP_775259.1 | 12 - 13 | 1833 | 61 | |
XM_017024234.1 | XP_016879723.1 | 11 - 12 | 1708 | 61 | |
GenBank mRNA | AF539794.1 | - | 12 - 13 | 1833 | 61 |
AL832355.1 | - | 13 - 14 | 1918 | 61 | |
BC101062.2 | - | 10 - 11 | 1590 | 61 | |
BC101063.2 | - | 12 - 13 | 1827 | 61 | |
BX647642.1 | - | 13 - 14 | 1924 | 61 |