The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135098.1 | NP_001128570.1 | 5 - 6 | 531 | 122 |
NM_173355.3 | NP_775491.1 | 3 - 4 | 534 | 122 | |
XM_005246359.3 | XP_005246416.1 | 3 - 4 | 534 | 122 | |
XM_017003484.1 | XP_016858973.1 | 3 - 4 | 534 | 122 | |
GenBank mRNA | AK122743.2 | - | 3 - 4 | 534 | 122 |
AY225131.1 | - | 3 - 4 | 359 | 122 | |
BC033529.2 | - | 5 - 6 | 531 | 122 | |
KU178873.1 | - | 3 - 4 | 340 | 122 | |
KU178874.1 | - | 3 - 4 | 340 | 122 |