Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC35F3
Entrez Gene ID
148641
Gene Name
solute carrier family 35 member F3
Gene Aliases
-
Chromosome Location
Chr.1: 233904712 - 234324518 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300845.1 NP_001287774.1 6 - 7 1313 67
NM_173508.3 NP_775779.1 7 - 8 1598 67
GenBank mRNA AK095031.1 - 7 - 8 1379 67
BC037878.1 - 6 - 7 1257 67

View Full Product Details