Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UMODL1
Entrez Gene ID
89766
Gene Name
uromodulin like 1
Gene Aliases
-
Chromosome Location
Chr.21: 42062959 - 42142996 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004416.2 NP_001004416.2 17 - 18 3143 58
NM_001199527.1 NP_001186456.1 16 - 17 3707 58
NM_001199528.2 NP_001186457.2 17 - 18 3323 58
NM_173568.3 NP_775839.3 16 - 17 3527 58
XM_011529797.1 XP_011528099.1 14 - 15 3135 58
XM_017028506.1 XP_016883995.1 16 - 17 3522 58
XM_017028507.1 XP_016883996.1 16 - 17 6451 58
XM_017028508.1 XP_016883997.1 6 - 7 1306 58
GenBank mRNA AB051810.1 - 16 - 17 3707 58
AB051811.1 - 17 - 18 3323 58
AK093654.1 - 2 - 3 385 58
AK124223.1 - 3 - 4 887 58
AY771619.1 - 16 - 17 3527 58
AY771620.1 - 17 - 18 3143 58

View Full Product Details