The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001200049.2 | NP_001186978.2 | 1 - 2 | 154 | 86 |
GenBank mRNA | AK098820.1 | - | 1 - 2 | 129 | 86 |
BC044661.1 | - | 1 - 2 | 112 | 86 | |
BX376477.2 | - | 1 - 2 | 154 | 86 | |
DR001134.1 | - | 1 - 2 | 392 | 86 |