The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173595.3 | NP_775866.2 | 6 - 7 | 639 | 93 |
XM_011538197.2 | XP_011536499.1 | 5 - 6 | 741 | 93 | |
XM_017019183.1 | XP_016874672.1 | 5 - 6 | 822 | 93 | |
XM_017019184.1 | XP_016874673.1 | 6 - 7 | 671 | 93 | |
GenBank mRNA | AB190990.1 | - | 6 - 7 | 656 | 93 |
AK125379.1 | - | 6 - 7 | 656 | 93 |