The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173611.3 | NP_775882.2 | 2 - 3 | 248 | 113 |
GenBank mRNA | AK095745.1 | - | 2 - 3 | 222 | 113 |
BC035600.1 | - | 1 - 2 | 106 | 113 | |
BC093898.1 | - | 2 - 3 | 218 | 113 | |
BP872319.1 | - | 2 - 3 | 248 | 113 | |
BP872410.1 | - | 2 - 3 | 248 | 113 |