The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166347.1 | NP_001159819.1 | 14 - 15 | 1729 | 73 |
NM_001166348.1 | NP_001159820.1 | 13 - 14 | 1659 | 73 | |
NM_001166349.1 | NP_001159821.1 | 13 - 14 | 1529 | 73 | |
NM_173626.3 | NP_775897.3 | 14 - 15 | 1726 | 73 | |
XM_006721833.1 | XP_006721896.1 | 14 - 15 | 1801 | 73 | |
XM_011524652.1 | XP_011522954.1 | 12 - 13 | 1562 | 73 | |
XM_011524653.1 | XP_011522955.1 | 12 - 13 | 1490 | 73 | |
XM_011524654.1 | XP_011522956.1 | 12 - 13 | 1375 | 73 | |
XM_017024505.1 | XP_016879994.1 | 13 - 14 | 1715 | 73 | |
XM_017024506.1 | XP_016879995.1 | 13 - 14 | 1715 | 73 | |
XM_017024507.1 | XP_016879996.1 | 14 - 15 | 1775 | 73 | |
GenBank mRNA | AF331524.1 | - | 6 - 7 | 503 | 73 |
AF345195.1 | - | 13 - 14 | 1461 | 73 | |
AJ544073.1 | - | 14 - 15 | 1700 | 73 | |
AK075248.1 | - | 14 - 15 | 1689 | 73 | |
AK315132.1 | - | 14 - 15 | 1692 | 73 | |
AL833468.1 | - | 13 - 14 | 1623 | 73 | |
BC035900.1 | - | 14 - 15 | 1668 | 73 | |
BC047451.1 | - | 8 - 9 | 778 | 73 |