Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARL10
Entrez Gene ID
285598
Gene Name
ADP ribosylation factor like GTPase 10
Gene Aliases
ARL10A
Chromosome Location
Chr.5: 176365468 - 176415085 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173664.5 NP_775935.1 3 - 4 658 126

View Full Product Details