The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013842.2 | NP_001013864.1 | 6 - 7 | 1106 | 69 |
NM_173686.2 | NP_775957.2 | 6 - 7 | 1082 | 69 | |
GenBank mRNA | AK092034.1 | - | 5 - 6 | 890 | 69 |
AK295405.1 | - | 6 - 7 | 1082 | 69 | |
BC012750.2 | - | 5 - 6 | 897 | 69 |