The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256063.1 | NP_001242992.1 | 4 - 5 | 452 | 90 |
NM_001256064.1 | NP_001242993.1 | 4 - 5 | 665 | 90 | |
NM_001843.3 | NP_001834.2 | 4 - 5 | 452 | 90 | |
NM_175038.2 | NP_778203.1 | 2 - 3 | 268 | 90 | |
XM_005268651.2 | XP_005268708.1 | 4 - 5 | 815 | 90 | |
XM_006719241.2 | XP_006719304.1 | 4 - 5 | 439 | 90 | |
XM_011537926.2 | XP_011536228.1 | 4 - 5 | 404 | 90 | |
XM_011537927.2 | XP_011536229.1 | 4 - 5 | 461 | 90 | |
XM_017018821.1 | XP_016874310.1 | 4 - 5 | 455 | 90 | |
XM_017018822.1 | XP_016874311.1 | 4 - 5 | 404 | 90 | |
XM_017018823.1 | XP_016874312.1 | 4 - 5 | 815 | 90 | |
XM_017018824.1 | XP_016874313.1 | 4 - 5 | 439 | 90 | |
XM_017018825.1 | XP_016874314.1 | 4 - 5 | 404 | 90 | |
XM_017018826.1 | XP_016874315.1 | 4 - 5 | 439 | 90 | |
XM_017018827.1 | XP_016874316.1 | 4 - 5 | 461 | 90 | |
GenBank mRNA | AK289544.1 | - | 4 - 5 | 544 | 90 |
AK289698.1 | - | 4 - 5 | 394 | 90 | |
BC036569.2 | - | 4 - 5 | 391 | 90 | |
U07819.1 | - | 3 - 4 | 225 | 90 | |
U07820.1 | - | 2 - 3 | 273 | 90 | |
Z21488.1 | - | 4 - 5 | 346 | 90 |