The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005076.3 | NP_005067.1 | 14 - 15 | 2087 | 82 |
XM_017002198.1 | XP_016857687.1 | 13 - 14 | 1828 | 82 | |
XM_017002199.1 | XP_016857688.1 | 15 - 16 | 2342 | 82 | |
GenBank mRNA | AK124736.1 | - | 9 - 10 | 1810 | 82 |
AK289985.1 | - | 13 - 14 | 1828 | 82 | |
BC129986.1 | - | 13 - 14 | 1853 | 82 | |
CR749495.1 | - | 13 - 14 | 2934 | 82 | |
X67734.1 | - | 14 - 15 | 1999 | 82 | |
X68274.1 | - | 14 - 15 | 1993 | 82 |