Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ST8SIA4
Entrez Gene ID
7903
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 4
Gene Aliases
PST, PST1, SIAT8D, ST8SIA-IV
Chromosome Location
Chr.5: 100806935 - 100903285 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005668.5 NP_005659.1 3 - 4 828 84
XM_005272078.2 XP_005272135.1 3 - 4 828 84
GenBank mRNA AK292872.1 - 3 - 4 780 84
BC053657.1 - 3 - 4 809 84
L41680.1 - 3 - 4 710 84

View Full Product Details