The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281437.1 | NP_001268366.1 | 4 - 5 | 579 | 82 |
NM_001281438.1 | NP_001268367.1 | 4 - 5 | 583 | 82 | |
NM_007002.3 | NP_008933.2 | 5 - 6 | 700 | 82 | |
NM_175573.2 | NP_783163.1 | 5 - 6 | 696 | 82 | |
XM_005260257.1 | XP_005260314.1 | 6 - 7 | 787 | 82 | |
XM_011528503.1 | XP_011526805.1 | 5 - 6 | 1535 | 82 | |
XM_017027602.1 | XP_016883091.1 | 5 - 6 | 1535 | 82 | |
XM_017027603.1 | XP_016883092.1 | 6 - 7 | 787 | 82 | |
XM_017027604.1 | XP_016883093.1 | 5 - 6 | 1531 | 82 | |
XM_017027605.1 | XP_016883094.1 | 5 - 6 | 1535 | 82 | |
XM_017027606.1 | XP_016883095.1 | 4 - 5 | 1418 | 82 | |
XM_017027607.1 | XP_016883096.1 | 4 - 5 | 1414 | 82 | |
XM_017027608.1 | XP_016883097.1 | 3 - 4 | 1204 | 82 | |
XM_017027609.1 | XP_016883098.1 | 3 - 4 | 1204 | 82 | |
XM_017027610.1 | XP_016883099.1 | 3 - 4 | 1204 | 82 | |
GenBank mRNA | AK307459.1 | - | - | 630 | 82 |
AL558522.3 | - | 5 - 6 | 632 | 82 | |
BC010733.1 | - | 2 - 3 | 212 | 82 | |
BC017245.2 | - | 5 - 6 | 596 | 82 | |
BG337785.1 | - | 4 - 5 | 538 | 81 | |
D64154.1 | - | 5 - 6 | 591 | 82 |