The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020453.3 | NP_065186.3 | 21 - 22 | 3922 | 115 |
XM_005248119.4 | XP_005248176.1 | 21 - 22 | 4008 | 115 | |
XM_005248120.4 | XP_005248177.1 | 20 - 21 | 3959 | 115 | |
XM_011513722.2 | XP_011512024.1 | 20 - 21 | 3858 | 115 | |
XM_017008472.1 | XP_016863961.1 | 22 - 23 | 4142 | 115 | |
GenBank mRNA | AB040920.1 | - | 10 - 11 | 1432 | 115 |
AJ441078.1 | - | 21 - 22 | 3922 | 115 | |
AK027598.1 | - | 14 - 15 | 2113 | 115 | |
AK074930.1 | - | 4 - 5 | 440 | 115 | |
BC131535.1 | - | 21 - 22 | 3900 | 115 |