Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BBS1
Entrez Gene ID
582
Gene Name
Bardet-Biedl syndrome 1
Gene Aliases
BBS2L2
Chromosome Location
Chr.11: 66510648 - 66533613 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024649.4 NP_078925.3 5 - 6 495 82
GenBank mRNA AF503941.1 - 5 - 6 495 82
AK095638.1 - 4 - 5 502 82
BC109064.2 - 5 - 6 488 82
BC109065.1 - 5 - 6 488 82
BX647612.1 - 5 - 6 524 82

View Full Product Details